Learn More
Ace Glass, Inc. 75mm ID 8" 75/50 epoxy coated flash chromatography column.
SDP
Supplier:  Ace Glass, Inc. 587233
Encompass_Preferred
75mm I.D., 8 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 75/50 joints.
Catalog No. 50-972-485