Learn More
Waters Corp Waters Corporation Phenyl Column, 90Å, 1.6 μm, 2.1 mm × 150 mm
Supplier:  Waters Corp 186008382
Encompass_Preferred
CORTECS Phenyl columns provide high efficiency and lower backpressures with complementary selectivity to C18 stationary phases. These columns ensure ultra-low MS bleed, long lifetimes, and excellent peak shape, making them suitable for various liquid chromatography applications.
  • 1.6 μm particles for exceptional separation efficiency.
  • 90Å pore size for good peak shape and resolution.
  • 2.1 mm inner diameter for high flow rates and enhanced productivity.
  • 150 mm length for cost-effectiveness and extended life.
  • Solid core technology for superior robustness.
  • End-capped to minimize MS bleed and boost analysis sensitivity.
  • Wide pH range of 2 to 8.
  • Suitable for analyzing peptides, proteins, small molecules, drugs, metabolites, and other biological compounds.
Catalog No. NC2025685