Learn More
Bio Rad Laboratories COLUMN AMINEX HPX-87N
Supplier:  Bio Rad Laboratories 1250143
Encompass
Columns, Carbohydrate Analysis; Bio-Rad; Aminex; HPX-87N; 300 x 7.8mm; Temperature: 85 deg.C; Flow rate: 0.6mL/min
Catalog No. 50-901-2146