Learn More
Trajan Scientific Column BPX35 QC2 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054711
Encompass_Preferred
0 22 mm ID x 0 25 m film thickness x 25 m length BPX35 GC capillary column
Catalog No. 50-004-94504