Learn More
Wilmad Labglass COLUMN CHROM 10.5X250MM 24/40
Supplier:  Wilmad Labglass LG4569T104
Encompass_Preferred
Column, Chromatography, w/ [ST] Joint, Fritted Disc and PTFE Stopcock. Column ID: 10.5mm; Column Length: 250mm; Joint Size: [ST] 24/40 Outer; Stopcock Size: [PS] 2. A sealed-in coarse porosity fritted disc.
Catalog No. 50-805-486