Learn More
Wilmad Labglass COLUMN CHROM 22X500MM 24/40
Supplier:  Wilmad Labglass LG4569T126
Encompass_Preferred
Column, Chromatography, w/ [ST] Joint, Fritted Disc and PTFE Stopcock. Column ID: 22mm; Column Length: 500mm; Joint Size: [ST] 24/40 Outer; Stopcock Size: [PS] 2. A sealed-in coarse porosity fritted disc.
Catalog No. 50-806-197
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.