Learn More
Wilmad Labglass COLUMN CHROM FRTD 10.5X250MM
Supplier:  Wilmad Labglass LG4567T104
Encompass_Preferred
Column, Chromatography, w/ Fritted Disc and PTFE Stopcock. Column ID: 10.5mm; Column Length: 250mm; Stopcock Size: [PS] 2. a sealed-in coarse porosity fritted disc to support the column packing.
Catalog No. 50-804-586