Learn More
Microsolv Technology Corporation Column Diamond Hydr 4.6X150Mm
Supplier:  Microsolv Technology Corporation 7000015P
Encompass_Preferred
HPLC Column, Diamond Hydride, 4um, 4.6mm ID x 150mm Length, 100A. Cogent TYPE-C Brand. 1/EA.
Catalog No. 50-014-51320