Learn More
Microsolv Technology Corporation Column Hplc Cholesterolcolumn
Supplier:  Microsolv Technology Corporation 6906905P2
Encompass_Preferred
HPLC Column, UDC Cholesterol, 4um, 2.1mm ID x 50mm Length, 100A. Cogent TYPE-C Brand. 1/EA.
Catalog No. 50-014-51293