Learn More
Waters Corp Column, HSS T3, 100Å, 1.8 μm, 2.1 mm X 150 mm
Supplier:  Waters Corp 180663540
Encompass_Preferred
This column features a universal, silica-based bonded phase compatible with 100% aqueous mobile phase, suitable for developing separations for polar and non-polar compounds. It exhibits ultra-low MS bleed and is scalable to HPLC separations, offering enhanced retention of polar compounds and metabolites using reversed-phase liquid chromatography without ion-pair reagents.
  • Achieves superior performance and polar compound retention with advanced T3 bonding.
  • Proprietary end-capping enhances performance, lifetime, peak shape, and stability.
  • Manufactured in a CGMP ISO 9001:2000 certified plant with ultra-pure reagents.
  • Tested chromatographically with various analytes to ensure reproducibility.
  • Designed to retain small, water-soluble, polar organic molecules.
  • Enables robust method development when combined with UPLC technology.
Catalog No. NC3947012