Learn More
Ace Glass, Inc. COLUMN PERF PLATE (5) 28MM PLT
SDP
Supplier:  Ace Glass, Inc. 656502
Encompass
Column, perforated plate,(5) 28mm plates,29/42 joints, 140mm effective length,245mm overall length
Catalog No. NC2283857