Learn More
LAMDA BIOTECH INC Column-Pure DNA Gel Recovery
Supplier:  LAMDA BIOTECH INC D507
Encompass_Preferred
A silica membrane-based spin column system designed for rapid purification of DNA fragments from agarose gels. Compatible with both TAE and TBE buffer systems for versatile DNA recovery applications.
Catalog No. NC3724895