Learn More
Applied Separations Sodium Sulfate SPE Column, 10 g Packing, 60 mL Cartridge Volume, 24 Pack
SDP
Supplier:  Applied Separations 14600
Encompass_Preferred
The Applied Separations Spe-ed SPE Sodium Sulfate Column is a solid phase extraction consumable designed for efficient sample preparation in laboratories. It features a 60 mL cartridge packed with 10 g of sodium sulfate, ensuring reliable drying and purification of samples. Supplied in a pack of 24, it enhances high-throughput workflows, making it essential for scientific applications.
Catalog No. NC2126649