Learn More
eMolecules​ 110-85-0 | Piperazine | Combi-Blocks | MFCD00005953 | 86.138 | C4H10N2 | 98.000 | C1CNCCN1 | 1kg | 335341891
Supplier:  eMolecules​ 335341891
Encompass_Preferred
Piperazine | Combi-Blocks | 110-85-0 | MFCD00005953 | 86.138 | C4H10N2 | 98.000 | C1CNCCN1 | 1kg | 335341891
Catalog No. 50-001-29368