Learn More
Sigma Aldrich Fine Chemicals Biosciences CombiTitrant 5 one-component reagent for volumetric Karl Fischer titration 1 ml ≙ ca. 5 mg H₂O Aquastar(R) |
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1880051000
Encompass
CombiTitrant 5 one-component reagent for volumetric Karl Fischer titration 1 ml ≙ ca. 5 mg H₂O Aquastar(R) |
Catalog No. NC1776051