Learn More
Medchemexpress LLC HY-N2146 5mg Medchemexpress, Combretastatin A4 CAS:117048-59-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21465MG
Encompass_Preferred
Medchemexpress, HY-N2146 5mg Combretastatin A4 CAS:117048-59-6 Combretastatin A4 is a microtubule -targeting agent that binds β-tubulin with K d of 0.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3890