Learn More
Bellco Glass, Inc. Compresion Reducer Fttg 1/2 - 1/4""
SDP
Supplier:  Bellco Glass, Inc. 196490135
Encompass_Preferred
These fittings are designed specifically for the bellco control systems and fit through a 5/16" fitting on the stainless steel headplate. MADE IN THE USA. Stainless steel Size: 1/2" – ¼"
Catalog No. 50-194-4937