Learn More
Research Products International Corp Compact Magnetic Stirrer, with Stainless Steel Plate
SDP
Supplier:  Research Products International Corp 410028
Encompass
These Compact Magnetic Stirrers are designed to provide years of rugged and dependable service in the laboratory environment.
Product Features:
Max. Stirring Quantity of H2O at 20L
Stainless Steel Work Plate Cover with Ceramic Material Provides Good Chemical Resistance Performance
Speed Range: 0-1500 rpm
Maintenance Free Brushless DC Motor
Catalog No. 50-999-095