Learn More
Yamato Scientific America, Inc. COMPACT STERILIZER 30L 115V
Supplier:  Yamato Scientific America, Inc. SK301C
Encompass_Preferred
Space-saving, 30L compact steam sterilizer, ideal for research facilities. Easy to use. Powerful 2000W pipe heater. Easy to read 4 digit LED display. Programmable sterilizing and temperature functions. Timer Setting Range 0 to 999 min. Increased safety features: Water level detection sensor with alarm, Overheat Protection, Sensor Lid closure sensor (Inter lock), Pressure Lamp Indicator. Temp setting range 50 to 126°C.
Catalog No. NC2261590