Learn More
Research Products International Corp Compact UV Lamp, 254 nm, 230 Volt
SDP
Supplier:  Research Products International Corp 353061230V
Encompass
Easy to handle and convenient to use. Lightweight lamps feature a durable plastic housing. Shortwave model, 254 nm, is equipped with long-life filter. Unit supplied with European cord for international use.
Catalog No. 50-136-7905