Learn More
ACROBiosystems Complement C2 25ug
Supplier:  ACROBiosystems CO2D82E525UG
Encompass_Preferred
Biotinylated Domestic pig Complement C2 Protein HisAvitag (CO2-D82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Leu 752 (Accession # A0A0B8RW16).
Catalog No. 50-253-5228