Learn More
Medchemexpress LLC Complement C3 protein, cynomolgus | >90.0% | 10 UG

Supplier: Medchemexpress LLC HYP7533210UG
Recombinant complement C3 protein from cynomolgus macaque, expressed in HEK293 cells with a C-terminal His tag. Supplied as a purified protein for complement pathway research, functional assays, and antibody or ligand binding studies. Purity is greater than 90% by reducing SDS-PAGE; follow recommended reconstitution and storage instructions for best activity.
- Purity greater than 90% as determined by reducing SDS-PAGE.
- Expressed in HEK293 cells to preserve mammalian post-translational modifications.
- C-terminal His tag for detection and purification.
- Available in small aliquots suitable for in vitro assays.
- Do not reconstitute below 100 μg/mL in ddH2O.
- Stable when frozen at -20°C; aliquot and store at -20°C or -80°C for long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.