Learn More
ABclonal Technology Complement C3 Rabbit PolymAb
SDP
Supplier:  ABclonal Technology A26625PM20UL
Encompass_Preferred
Complement component C3 plays a central role in the activation of complement system Its activation is required for both classical and alternative complement activation pathways The encoded preproprotein is proteolytically processed to generate alpha and beta subunits that form the mature protein which is then further processed to generate numerous peptide products The C3a peptide also known as the C3a anaphylatoxin modulates inflammation and possesses antimicrobial activity Mutations in this gene are associated with atypical hemolytic uremic syndrome and age-related macular degeneration in human patients
Catalog No. 50-000-11956