Learn More
Carbosynth LLC. Purified Complement C3d protein isolated from human plasma
SDP
Supplier:  Carbosynth LLC. 32CCP10430.1MG
Encompass_Preferred
C3d plays a role in enhancing the B-cell response and is produced during the classical complement activation pathway following the proteolytic cleavage of C3b by Factor I.
Catalog No. 50-125-3294