Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Complement C5-in-1 | 2365402-67-9 | 99.1% | 444.52 g/mol | C24H32N2O6 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1283425MG

Encompass_Preferred

Complement C5-IN-1 is a small-molecule inhibitor of complement component C5 designed for research use in complement biology and immunology. Supplied as a solid, it is characterized by documented purity, molecular properties, solubility data, and recommended storage conditions to support experimental reproducibility.

  • Inhibits complement component C5 to prevent its cleavage.
  • High purity (99.14%) suitable for biochemical assays.
  • Molecular weight 444.52 g/mol; formula C24H32N2O6.
  • Soluble in DMSO (≥100 mg/mL) with in vivo vehicle options provided.
  • Stable as a powder when stored at -20 °C for long-term storage.

Catalog No. 50-002-64344


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.