Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Complement component C3 (cynomolgus) | >90.0% | 100 UG
SDP

Supplier:  Medchemexpress LLC HYP75332100UG

Encompass_Preferred

Recombinant complement component C3 derived from cynomolgus (Macaca fascicularis), expressed in HEK293 cells and C-terminally His-tagged. Supplied lyophilized from 0.22 μm filtered PBS (pH 7.4), the product is intended for research use in complement system and immunology studies and is available in multiple pack sizes. Store at -20°C; after reconstitution it is stable at 4°C for 1 week or at -20°C for longer-term storage with carrier protein.

  • Recombinant cynomolgus origin (Macaca fascicularis).
  • Expressed in HEK293 mammalian cells.
  • C-terminal His tag for detection and purification.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Lyophilized formulation from PBS, pH 7.4.
  • Available in multiple pack sizes including 100 μg.
  • Recommend reconstitution to not less than 100 μg/mL.

Catalog No. 50-002-91107


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.