Learn More
Medchemexpress LLC Complement factor D-IN-1 | 1386455-76-0 | 98.9% | 483.32 g/mol | C21H19BrN6O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10203425MG
Encompass_Preferred
Complement factor D-IN-1 is a potent and selective reversible small-molecule inhibitor of complement factor D used in research to probe the alternative complement pathway. It exhibits low-nanomolar activity in biochemical assays and is supplied with high reported purity as a solid or as a DMSO solution.
  • Potent and selective reversible inhibitor of complement factor D.
  • Demonstrated low-nanomolar IC50s in FD thioesterolytic and MAC deposition assays.
  • High reported purity (≈98.8%).
  • Available as dry powder or as a ready-to-use DMSO solution.
  • Stable when stored under recommended conditions for long-term use.
  • Suitable for biochemical and cellular complement pathway studies.
Catalog No. 50-002-47483