Learn More
Medchemexpress LLC Complement factor D-IN-2 | 1903742-34-6 | 99.3% | 574.43 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY13828110MM/1ML
Encompass_Preferred
Complement factor D-IN-2 is an inhibitor of complement factor D, derived from patent WO2015130838A1, compound 190. It specifically targets factor D, thereby inhibiting the complement cascade at an early and crucial stage of the alternative complement pathway. This compound is suitable for research on autoimmune diseases and has been shown to reduce the excessive activation of complement factor D.
  • Inhibits complement factor D
  • Targets factor D to inhibit the complement cascade
  • Useful for research on autoimmune diseases
  • Reduces excessive activation of complement factor D
Catalog No. 50-003-50110