Learn More
Cell Biologics Complete Human Epithelial Cell Medium /W Kit - 500 mL
SDP
Supplier:  Cell Biologics H6621
Encompass_Preferred
Human Epithelial Cell Medium is a complete medium designed for the culture of human and monkey epithelial cells. It was tested and optimized with epithelial cell growth and proliferation in vitro.  It is formulated for use with 5% CO2 and 95% air in a humidified incubator.
Catalog No. NC1193234