Learn More
Medchemexpress LLC Compound 134a | 2636840-37-2 | 98.4% | 566.58 g/mol | C29H32F6N4O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14840410MG
Encompass_Preferred
p53 Activator 5 is a small-molecule research compound that binds mutant p53 and restores its DNA-binding and transcriptional activity, displaying anti-tumor effects in preclinical studies. It is intended for use as a high-purity reagent in in vitro and preclinical investigations of p53-mediated apoptosis and related signaling pathways.
  • Restores mutant p53 DNA-binding activity.
  • Demonstrates anti-tumor activity in preclinical models.
  • Acts as a p53 pathway activator, promoting apoptosis.
  • Suitable as a solid research reagent for in vitro studies.
  • High purity (98.4%) for reliable experimental results.
  • Molecular weight 566.58 g/mol; formula C29H32F6N4O.
Catalog No. 50-002-86024