Learn More
Medchemexpress LLC Compound 14a | 1198098-03-1 | 98.0% | C12H12N2O3S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14713410MG
Encompass_Preferred
GSK-3β inhibitor 10 is a small-molecule research compound that selectively inhibits glycogen synthase kinase-3β. It is a potent inhibitor (reported IC50 ≈ 80 nM) used in preclinical studies of Alzheimer's disease and PI3K/Akt/mTOR signaling. The compound has formula C12H12N2O3S, molecular weight 264.3, and product purity listed as ≈98%.
  • Potent GSK-3β inhibition (reported IC50 ≈ 80 nM).
  • Suitable for preclinical biochemical and cellular research.
  • High purity for research use (≈98%).
  • Well-characterized chemical identity: CAS 1198098-03-1, formula C12H12N2O3S.
  • Molecular weight 264.3 for calculation of dosing and solution preparation.
Catalog No. 50-002-58880