Learn More
Selleck Chemical LLC Compound 32-E1407-1MG
SDP
Supplier:  Selleck Chemical LLC E14071MG
Encompass_Preferred
-Galactosylceramide ( -GalCer) is a synthetic glycolipid with antitumorial and immunostimulatory -Galactosylceramide is a very potent NKT cell agonist and binds effectively to CD1d The complex of -Galactosylceramide plus CD1d binds the NKT cell TCR (T cell antigen receptor)
Catalog No. 50-313-6584