Learn More
Medchemexpress LLC Compound 48/80 (trimer) | 94724-12-6 | 95.0% | 519.72 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY115768A
Encompass_Preferred
Compound 48/80 (trimer) is a trimer of Compound 48/80. It is widely used in animal and tissue models as a selective mast cell activator, making it a valuable tool in biological research.
  • A trimer of Compound 48/80.
  • Used in animal models.
  • Used in tissue models.
  • Acts as a selective mast cell activator.
Catalog No. 50-004-39058