Learn More
TARGETMOL CHEMICALS INC COMPOUND C108 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T383875MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Compound C108 is an inhibitor of GTPase-activating protein SH3 domain-binding protein 2 (G3BP2). Compound C108 can be used in studies about breast tumors and esophageal squamous cell carcinoma. purity: 99%
Catalog No. 50-258-0107