Learn More
STEMCELL Technologies Compound E, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 73954
Encompass
Compound E is a potent, cell-permeable, selective inhibitor that blocks the cleavage of both gamma-secretase and the Notch intracellular domain with similar IC50 values, ranging from 0.24 to 0.37 nM (Beher et al.; Seiffert et al.). DIFFERENTIATION· Inhibits growth, differentiation, and motility of neuroblastoma cells (Ferrari-Toninelli et al.).· Accelerates the differentiation of human embryonic stem cells into primitive neural stem cells (Li et al.).
Catalog No. NC1507417
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov