Learn More
Chemglass Life Sciences PFA 90° Adapter, 1/8" Compression to 1/4" NPT
SDP
Supplier:  Chemglass Life Sciences CG1975E16
Encompass_Preferred
PFA 90¿ Adapter, 1/8" Compression to 1/4" NPT. PFA 90¿ adapter, compression to male NPT fittings are ideally suited for use in corrosive environments and chemical applications. The fitting design yields a leak-tight connection and requires no special tools to assemble. The connection is achieved by merely inserting the tubing into the fitting assembly and tightening the nut.
Catalog No. 50-143-595