Learn More
Crystal Chem Inc Control for the Rat Total Bile Acid (Enzyme Cycling) Kit
SDP
Supplier:  Crystal Chem Inc 80363
Encompass
The Control for the Rat Total Bile Acid Assay (Enzyme Cycling) can be used to validate the calibration curve obtained when running the Rat Total Bile Acid Assay (Enzyme Cycling). Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7699
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.