Learn More
Apexbio Technology LLC Concanavalin A (Con A)-Biotin 5mg(5 mg/mL)
SDP
Supplier:  Apexbio Technology LLC K441225
Encompass_Preferred
Concanavalin A (Con A) is a lectin protein derived from Canavalia ensiformis (jack bean) with a molecular weight of approximately 104 kDa Each subunit can bind one Ca2 and one Mn2 ion and contains a carbohydrate-binding site Upon binding these metal ions Con A specifically binds to glycoproteins glycolipids and carbohydrates containing -D-glucose and -D-mannose residues Biotin is a small-molecule water-soluble vitamin that exhibits very strong affinity to avidin Concanavalin A-Biotin is a labeled protein conjugate composed of Concanavalin A linked to biotin widely used in biological research especially for detecting and quantifying biomolecules containing specific carbohydrate structures through immunohistochemistry (IHC) immunocytochemistry (ICC) and enzyme-linked immunosorbent assay (ELISA)
Catalog No. 50-310-7106