Learn More
Apexbio Technology LLC Concanavalin A (Con A)-HyperFluor 647 5mg(5 mg/mL)
SDP
Supplier:  Apexbio Technology LLC K441625
Encompass_Preferred
Concanavalin A (Con A) derived from Canavalia ensiformis (jack bean) is a plant lectin protein with a molecular weight of approximately 104 kDa Each subunit can bind one Ca2 ion and one Mn2 ion and has one carbohydrate binding site Con A is capable of binding to various glycoproteins glycolipids and -D-glucose and -D-mannose residues HyperFluorTM 647 is a fluorescent dye that emits green fluorescence under UV light (Ex/Em 656 nm/670 nm) This conjugation provides Con A-HyperFluorTM 647 a high visualization capability when detecting carbohydrate molecules on cell surfaces This product is HyperFluorTM 647-labeled Concanavalin A provided in solution form It is applicable in glycobiology research serving as a probe to detect carbohydrate molecules on the cell surface and analyzed effectively using flow cytometry Applying IHC staining procedures with this product allows visualization of carbohydrate distribution in tissues or cells under a fluorescence microscope
Catalog No. 50-310-7011