Learn More
Medchemexpress LLC Concanamycin A | 80890-47-7 | 866.09 | 50 UG
SDP
Supplier:  Medchemexpress LLC HYN172450UG
Encompass_Preferred
Concanamycin A, also known as Folimycin or Antibiotic X 4357B, is a macrolide antibiotic and a potent vacuolar type H+-ATPase (V-ATPase) inhibitor. It also effectively inhibits lysosomal acidification and has applications in T cell-mediated inflammation research. This product originates from S. diastatochromogenes as a microbial metabolite with immunomodulatory activity.
  • Macrolide antibiotic.
  • Vacuolar type H+-ATPase (V-ATPase) inhibitor.
  • Inhibitor of lysosomal acidification.
  • Used in T cell-mediated inflammation research.
  • Originates from S. diastatochromogenes.
  • Available in solid form, white to off-white in color.
Catalog No. 50-004-16308