Learn More
Abcam Concanamycin B, H+-ATPase inhibitor, 100UG
Supplier:  Abcam AB144228100UG
Encompass_Preferred
MW 852.1 Da, Purity >98%. Potent, selective vacuolar type H+-ATPase inhibitor. Antibiotic agent. Induces EGF-dependent apoptosis. Shows similar effects as Concanamycin A (ab144227) and Concanamycin C (ab144229). Shows potent antiviral effects (IC50 = 6 nM, HSV-1 replication).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5970