Learn More
Abcam Concanamycin C, H+-ATPase inhibitor, 500UG
Supplier:  Abcam AB144229500UG
Encompass_Preferred
MW 823.1 Da, Purity >98%. Potent, selective vacuolar-type H+-ATPase inhibitor. Macrolide antibiotic agent. Concanamycin A (ab144227) analog. Shows antiproliferative and potent antifungal effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5965