Learn More
AdipoGen Concanamycin C high purity
SDP
Supplier:  AdipoGen BVT0254C100
Encompass_Preferred
Chemical. CAS 81552-34-3. Formula C45H74O13. MW 823.1. Isolated from Streptomyces sp. Antibiotic. Vacuolar-type H-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Anti-osteoporotic. Antiviral. Immunosuppressive. Antifungal and anti-yeast.
Catalog No. 50-256-2039