Learn More
Cayman Chemical ConcnamycIn A 100ug
SDP
Supplier:  Cayman Chemical 11050100 UG
Encompass_Preferred
A plecomacrolide antibiotic; active against S. cerevisiae, S. sake, A. citri, P. citrinum, and P. oryzae (MICs = 50 = ~2.1-2.3 μM); decreases perforin levels in CD8+ OE4 CTLs; inhibits concanavalin A-induced thymidine incorporation by 99% in isolated mouse splenic lymphocytes at 100 μM; prevents influenza virus entry into MDCK cells at 0.8 μM
Catalog No. 50-295-4814