Learn More
Grainger Concrete Chain Saw Chain 16 Chain L
Supplier:  Grainger 14H223
Encompass_Preferred
The ICS Concrete Chain Saw Chain For 26KR18 is a concrete cutting chain designed for the 26KR18 concrete saw. It features a .404" pitch, .063" gauge, and 18" bar length. The chain is constructed with heat-treated chrome alloy steel for durability and long life. It is designed for fast, efficient concrete cutting with minimal vibration. The chain can be purchased individually or as part of a kit that includes the bar, chain, and drive links. Proper chain tensioning and bar/chain lubrication are recommended for optimal performance. This heavy-duty concrete cutting chain is an ideal replacement part for the 26KR18 concrete saw.
Catalog No. 50-275-0675