Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wilmad Labglass CONDENSER AIR 750MM 24/40

Supplier:  Wilmad Labglass LG4670106

Encompass_Preferred

Air Condenser 750mm 24/40 (Encompass)

Catalog No. 50-803-831

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.