Learn More
Wilmad Labglass CONDENSER ALLIHN 24/40 600MM
Supplier:  Wilmad Labglass LG4680110
Encompass_Preferred
Condenser, Allihn. Jacket Length: 600mm; Joint Size: [ST] 24/40. Allihn Condenser w/ a standard taper Outer joint at the top and a standard taper inner drip tip joint of the same size at the bottom.
Catalog No. 50-803-628
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.