Learn More
Wilmad Labglass CONDENSER ALLIHN SXHLT 45/50
Supplier:  Wilmad Labglass LG6900114
Encompass_Preferred
Condenser, Allihn, For Soxhlet Extraction Apparatus. Joint Size: 45/50. Replacement component For LG-6900-104 Soxhlet Extraction Apparatus.
Catalog No. 50-803-524