Learn More
Wilmad Labglass CONDENSER GRAHAM 24/40 400MM
Supplier:  Wilmad Labglass LG4940106
Encompass_Preferred
Condenser, Graham. Jacket Length: 400mm; Joint Size: 24/40. Condenser has standard taper Outer joint at the top and standard taper inner drip tip joint of the same size at the bottom.
Catalog No. 50-803-853
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.