Learn More
Wilmad Labglass CONDENSER WEST 24/40 250MM
Supplier:  Wilmad Labglass LG5350102
Encompass_Preferred
Condenser, West. Jacket Length: 250mm; Joint Size: [ST] 24/40. w/ a standard taper Outer joint at the top and a standard taper inner drip tip joint of the same size at the bottom.
Catalog No. 50-803-435
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.